
Eclalbasaponin I

Synonym(s):
(3β,16α)-3-(β-D-Glucopyranosyloxy)-16-hydroxyolean-12-en-28-oic acid β-D-glucopyranosyl ester
Empirical Formula (Hill Notation):
C42H68O14
CAS Number:
Molecular Weight:
796.98
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing