
Donepezil-N-oxide

Synonym(s):
2,3-Dihydro-5,6-dimethoxy-2-[[1-oxido-1-(phenylmethyl)-4-piperidinyl]methyl]- 1H-inden-1-one
Empirical Formula (Hill Notation):
C24H29NO4
CAS Number:
Molecular Weight:
395.49
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing