
Dihydrokavain

Synonym(s):
(S)-(+)-7,8-Dihydrokavain, (6S)-5,6-Dihydro-4-methoxy-6-(2-phenylethyl)-2H-pyran-2-one, 7,8-Dihydrokawain, Dihydrokavain, Marindinin
Empirical Formula (Hill Notation):
C14H16O3
CAS Number:
Molecular Weight:
232.28
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing