
β-Sitosterol β-D-glucoside

Synonym(s):
β-Daucosterol, Coriandrinol, Daucosterin, Eleutheroside A
Empirical Formula (Hill Notation):
C35H60O6
CAS Number:
Molecular Weight:
576.85
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing