
Beclometasone dipropionate for peak identification

Synonym(s):
Beclomethasone dipropionate, 9-Chloro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17,21-dipropionate
Empirical Formula (Hill Notation):
C28H37ClO7
CAS Number:
Molecular Weight:
521.04
Products
Formula Weight
Gene Alias
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing