
Baccatin III

Synonym(s):
O-De[(2R,3S)-3-(Benzoylamino)-2-hydroxy-3-phenylpropanoyl]paclitaxel
Empirical Formula (Hill Notation):
C31H38O11
CAS Number:
Molecular Weight:
586.63
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3