
α-Tocotrienol

Synonym(s):
D-α-Tocotrienol, (R)-α-Tocotrienol, [R-(E,E)]-3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyl-3,7,11-tridecatrienyl)-2H-1-benzopyran-6-ol
Empirical Formula (Hill Notation):
C29H44O2
CAS Number:
Molecular Weight:
424.66
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing