Skip to Content
Merck
4-tert-Butylbenzoic acid 99%

4-tert-Butylbenzoic acid

4-tert-Butylbenzoic acid 99%
Synonym(s):
4-tert-Butylbenzoic acid
Linear Formula:
4-[(CH3)3C]C6H4COOH
CAS Number:
Molecular Weight:
178.23

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing