Merck
5.09227
G 418 Sulfate, Sterile-Filtered Solution in PBS, Cell Culture-Tested
Synonym(s):
G 418 Sulfate, Sterile-Filtered Solution in PBS, Cell Culture-Tested, G418, G 418
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
UNSPSC Code:
12352203
MDL number:
Technical Service
Let Us Assist
assay
≥98% (TLC)
Quality Segment
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze, avoid repeated freeze/thaw cycles, protect from light
storage temp.
2-8°C
InChI
1S/C20H40N4O10.2H2O4S/c1-6(25)14-11(27)10(26)9(23)18(32-14)33-15-7(21)4-8(22)16(12(15)28)34-19-13(29)17(24-3)20(2,30)5-31-19;2*1-5(2,3)4/h6-19,24-30H,4-5,21-23H2,1-3H3;2*(H2,1,2,3,4)/t6?,7-,8+,9+,10+,11-,12-,13-,14+,15+,16-,17-,18+,19-,20+;;/m0../s1
InChI key
UHEPSJJJMTWUCP-NKCAIAFTSA-N
General description
The optimal concentration of G418 for selection of resistance will vary according to the organism and/or cell type under investigation. In general, the concentration of active drug required for selection is as follows: Dictyostelium sp.: 10-100 µg/ml Plant cells: 10-100 µg/ml Yeast cells: 0.5-1.0 µg/ml Mammalian cells: 0.1-2.0 µg/ml
Packaging
Packaged under inert gas
Physical form
Sterile-filtered solution of Cat. No. 345810 supplied at 50 mg/ml in PBS, pH 7.
Preparation Note
Following initial thaw, aliquot and freeze (-20°C). Aliquots are stable for up to 1 year at -20°C.
Other Notes
Ethier, S.P., and Taback, E. 1993. Cancer Lett.74, 189.
Santerre, R.F., et al. 1991. Methods Mol. Biol.7, 245.
Maniatis, T., et al. 1989. In Molecular Cloning, A Laboratory Manual, Second Edition, Cold Spring Harbor, NY.
Edwards, S.A., and Adamson, E.D. 1987. J. Cell Physiol.133, 46.
Ernst, J.F., and Chan, R.K. 1985. J. Bacteriol.163, 8.
Canaani, D., and Berg, P. 1982. Proc. Natl. Acad. Sci. USA79, 5166.
Hirth, K.P., et al. 1982. Proc. Natl. Acad. Sci. USA79, 7356.
Ursic, D., et al. 1981. Biochem. Biophys. Res. Commun.101, 1031.
Jimenez, A., and Davies, J. 1980. Nature287, 869.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
pictograms
Health hazard
signalword
Danger
Hazard Classifications
Resp. Sens. 1 - Skin Sens. 1
Storage Class
12 - Non Combustible Liquids
wgk
WGK 3
flash_point_c
Not applicable
flash_point_f
Not applicable
hcodes