Merck
A cleavable amino-thiol linker for reversible linking of amines to DNA.
The Journal of organic chemistry (2006-12-02)
Jonas W Højfeldt, Peter Blakskjaer, Kurt V Gothelf
ABSTRACT
A cleavable heterobifunctional cross-linker for the reversible conjugation of amines to thiol-modified DNA has been developed and tested. The succinimidyl 2-(vinylsulfonyl)ethyl carbonate (SVEC) was prepared in three steps and tested for its ability to react with amines and thiols. The linker was efficient for binding leucine to a thiol-modified DNA sequence and for releasing the amino acid at pH 11.8.
Sigma-Aldrich
N,N′-Disuccinimidyl carbonate, ≥95%
Sigma-Aldrich
4-Nitrophenyl chloroformate, 96%