Merck
  • Perchlorate levels in samples of sodium nitrate fertilizer derived from Chilean caliche.
Perchlorate levels in samples of sodium nitrate fertilizer derived from Chilean caliche.
Environmental pollution (Barking, Essex : 1987) (2001-04-09)
E T Urbansky, S K Brown, M L Magnuson, C A Kelty
ABSTRACT
Paleogeochemical deposits in northern Chile are a rich source of naturally occurring sodium nitrate (Chile saltpeter). These ores are mined to isolate NaNO3 (16-0-0) for use as fertilizer. Coincidentally, these very same deposits are a natural source of perchlorate anion (ClO4-). At sufficiently high concentrations, perchlorate interferes with iodide uptake in the thyroid gland and has been used medicinally for this purpose. In 1997, perchlorate contamination was discovered in a number of US water supplies, including Lake Mead and the Colorado River. Subsequently, the Environmental Protection Agency added this species to the Contaminant Candidate List for drinking water and will begin assessing occurrence via the Unregulated Contaminants Monitoring Rule in 2001. Effective risk assessment requires characterizing possible sources, including fertilizer. Samples were analyzed by ion chromatography and confirmed by complexation electrospray ionization mass spectrometry. Within a lot, distribution of perchlorate is nearly homogeneous, presumably due to the manufacturing process. Two different lots we analyzed differed by 15%, containing an average of either 1.5 or 1.8 mg g-1. Inadequate sample size can lead to incorrect estimations; 100-g samples gave sufficiently consistent and reproducible results. At present, information on natural attenuation, plant uptake, use/application, and dilution is too limited to evaluate the significance of these findings, and further research is needed in these areas.
Sigma-Aldrich
Sodium nitrate, ACS reagent, ≥99.0%