- Moderating black powder chemistry for the synthesis of doped and highly porous graphene nanoplatelets and their use in electrocatalysis.
Moderating black powder chemistry for the synthesis of doped and highly porous graphene nanoplatelets and their use in electrocatalysis.
Inspired by black powder explosion, a mixture of sugar and nitrate is diluted with inert molten salt, such that their rapid reaction is moderated, allowing capturing of the intermediacy product: highly porous N-doped graphene nanoplates. The methodology is extended to S-doped porous carbon sheets by replacing the nitrate with sulfate. These MS-derived carbons show promising performances towards electrocatalysis and capacitive energy storage.