Merck
  • Complications following sedation of paediatric oncology patients undergoing radiotherapy.
Complications following sedation of paediatric oncology patients undergoing radiotherapy.
West African journal of medicine (1999-01-28)
A Adenipekun, O A Soyannwo, S D Amanor-Boadu, O B Campbell, A R Oyesegun
ABSTRACT
Sedation is often required to achieve immobilisation of small children during radiotherapy to avoid irradiation of normal tissues during the course of treatment. At the University College Hospital, Ibadan radiotherapists provide sedation for such patients with administration of parenteral and/or oral promethazine, diazepam, chlorpromazine and paraldehyde. This retrospective review of 84 children aged 1 month to 6 years who received sedation for radiotherapy over a period of twenty-one to twenty-eight days showed that 48% had complications. These included injection cellulitis (85.3%), injection abscess (4.87%), paresis of the lower limb (7.3%), aspiration pneumonia (2.4%). Anaesthetists in developing countries should be encouraged to extend their expertise in caring and resuscitation of sedated or unconscious patients to the radiotherapy unit. This will allow for the use of a wider variety of sedative agents and better monitoring as well as minimise or eradicate complications.
Sigma-Aldrich
Paraldehyde, ≥97%
Sigma-Aldrich
Paraldehyde, ≥97.0% (GC)
Sigma-Aldrich
Paraldehyde, Arxada quality, ≥99.5% (GC)