Merck
  • Restoration of the activity of active-site mutants of the hyperthermophilic beta-glycosidase from Sulfolobus solfataricus: dependence of the mechanism on the action of external nucleophiles.
Restoration of the activity of active-site mutants of the hyperthermophilic beta-glycosidase from Sulfolobus solfataricus: dependence of the mechanism on the action of external nucleophiles.
Biochemistry (1998-12-23)
M Moracci, A Trincone, G Perugino, M Ciaramella, M Rossi
ABSTRACT
The beta-glycosidase from the hyperthermophilic Archaeon Sulfolobus solfataricus hydrolyzes beta-glycosides following a retaining mechanism based upon the action of two amino acids: Glu387, which acts as the nucleophile of the reaction, and Glu206, which acts as the general acid/base catalyst. The activities of inactive mutants of the catalytic nucleophile Glu387Ala/Gly were restored by externally added nucleophiles. Sodium azide and sodium formate were used as external nucleophiles and the products of their reaction were characterized. Glu387Ala/Gly mutants were reactivated with 2, 4-DNP-beta-Glc substrate and the Glu387Gly mutant showed recovered activity, with the same nucleophiles, also on 2-NP-beta-Glc. The reaction catalyzed by the Glu387Gly mutant proceeded differently depending on the type of externally added nucleophile. Sodium azide restored the catalytic activity of the mutant by attacking the alpha-side of the anomeric carbon of the substrates, thereby yielding an inverting glycosidase. Sodium formate promoted the opposite behavior (retaining) in the mutant, producing 3-O-beta-linked disaccharide derivative of the substrates. A possible role of sodium formate as a biomimicking agent in replacing the natural nucleophile Glu387 is also discussed.
Sigma-Aldrich
2-Nitrophenyl β-D-glucopyranoside, powder
Pricing and availability is not currently available.