- Detection of inactive precursors of beta-glucanases in Saccharomyces cerevisiae.
Detection of inactive precursors of beta-glucanases in Saccharomyces cerevisiae.
FEBS letters (1983-09-19)
L M Hernandez, I Olivero, G Larriba
PMID6413252
ABSTRACT
Accumulation and secretion of beta-glucanases have been studied in vivo by using a thermosensitive secretory mutant of Saccharomyces cerevisiae blocked at the endoplasmic reticulum level (sec 18-1). When incubated at the restrictive temperature no accumulation of active glucanases was observed. Following a shift to permissive conditions in the presence of cycloheximide a rise in the internal activity took place. The increase in total glucanase activity was partially due to the activation of an exo-glucanase that hydrolyzes PNPG. It is concluded that glucanases are synthesized in inactive precursor forms and are converted to the active forms in their secretory pathway.