Merck
  • Early uptake and continuous accumulation of thallium-201 chloride in a benign mixed tumor of soft tissue: case report.
Early uptake and continuous accumulation of thallium-201 chloride in a benign mixed tumor of soft tissue: case report.
Diagnostic pathology (2010-06-01)
Shigenori Nagata, Yu-Fen Jin, Katsuhiko Yoshizato, Masanori Kitamura, Norishige Iizuka, Misa Song, Miki Tomoeda, Michiko Yuki, Chiaki Kubo, Hidenori Yoshizawa, Hidetatsu Outani, Kenichiro Hamada, Nobuhito Araki, Masahiro Funauchi, Yasuhiko Tomita
ABSTRACT
A case of benign mixed tumor of the soft tissue in a 64-year-old Japanese male is presented. He noticed a painless, elastic hard mass sized 3 cm in the right knee, which gradually grew larger and harder in the last 5 years. Magnetic resonance imaging demonstrated a mass lesion embedded in the subcutaneous tissue with low and high signal intensity at T1- and T2-weighted images, respectively. Tl-201 scintigraphy showed an early uptake of Tl-201 within the lesion at 10 minutes after injection, which was slightly decreased but still continued at 2 hours later. The patient underwent a resection of tumor, and the pathological diagnosis was a benign mixed tumor of soft tissue without high vascularity, characterized by histological features similar to pleomorphic adenomas in the salivary glands. Immunohistochemical study proved expression of Na+/K+-ATPase of tumor cells. Overexpression of Na+/K+-ATPase of the tumor might be responsible for the early uptake of Tl-201, and poor vascular structure in this tumor might lead to continuous accumulation. The Tl-201 scintigraphic features of mixed tumor of soft tissue are assessed to resemble those of malignant soft tissue tumors.
Sigma-Aldrich
Thallium(I) chloride, 99%
Sigma-Aldrich
Thallium(I) chloride, 99.999% trace metals basis