Merck
  • Towards a molecular genetic system for the pathogenic fungus Paracoccidioides brasiliensis.
Towards a molecular genetic system for the pathogenic fungus Paracoccidioides brasiliensis.
Fungal genetics and biology : FG & B (2007-05-22)
A J Almeida, J A Carmona, C Cunha, A Carvalho, C A Rappleye, W E Goldman, P J Hooykaas, C Leão, P Ludovico, F Rodrigues
ABSTRACT
We herein report the development of a molecular toolbox for the dimorphic fungus Paracoccidioides brasiliensis, specifically a more efficient transformation and a gene expression system. We evaluated several parameters that influence Agrobacterium tumefaciens-mediated transformation (ATMT), such as co-cultivation conditions and host cell susceptibility. Our results show that cellular recovery and air drying of A. tumefaciens:P. brasiliensis mixtures are essential for ATMT. Overall, our data indicate a transformation efficiency of 78+/-9 transformants/co-cultivation (5+/-1 transformants/10(6) target cells). P. brasiliensis GFP-expressing isolates were also constructed by insertion of the GFP gene under the control of several fungal promoters. RT-PCR, epifluorescence microscopy and flow cytometry analysis revealed Gfp visualization for all studied promoters but without significant differences in fluorescence and gene expression levels. Moreover, we present evidence for the occurrence of random single gene copy integration per haploid nuclei and the generation of homokaryon progeny, relevant for the future use in targeted mutagenesis and linking mutations to phenotypes.
Sigma-Aldrich
Azaserine, Hybri-Max, γ-irradiated, 50x, lyophilized powder, BioXtra, suitable for hybridoma
Sigma-Aldrich
Azaserine, ≥98% (TLC)