Merck
  • The polypyrimidine tract binding protein binds upstream of neural cell-specific c-src exon N1 to repress the splicing of the intron downstream.
The polypyrimidine tract binding protein binds upstream of neural cell-specific c-src exon N1 to repress the splicing of the intron downstream.
Molecular and cellular biology (1997-08-01)
R C Chan, D L Black
ABSTRACT
The neural cell-specific N1 exon of the c-src pre-mRNA is both negatively regulated in nonneural cells and positively regulated in neurons. We previously identified conserved intronic elements flanking N1 that direct the repression of N1 splicing in a nonneural HeLa cell extract. The upstream repressor elements are located within the polypyrimidine tract of the N1 exon 3' splice site. A short RNA containing this 3' splice site sequence can sequester trans-acting factors in the HeLa extract to allow splicing of N1. We now show that these upstream repressor elements specifically interact with the polypyrimidine tract binding protein (PTB). Mutations in the polypyrimidine tract reduce both PTB binding and the ability of the competitor RNA to derepress splicing. Moreover, purified PTB protein restores the repression of N1 splicing in an extract derepressed by a competitor RNA. In this system, the PTB protein is acting across the N1 exon to regulate the splicing of N1 to the downstream exon 4. This mechanism is in contrast to other cases of splicing regulation by PTB, in which the protein represses the splice site to which it binds.
Sigma-Aldrich
SRC N1 human, ≥60% (SDS-PAGE), recombinant, expressed in baculovirus infected insect cells, buffered aqueous glycerol solution