Merck
  • Activities of lectins and their immobilized derivatives in detergent solutions. Implications on the use of lectin affinity chromatography for the purification of membrane glycoproteins.
Activities of lectins and their immobilized derivatives in detergent solutions. Implications on the use of lectin affinity chromatography for the purification of membrane glycoproteins.
Biochemistry (1977-05-03)
R Lotan, G Beattie, W Hubbell, G L Nicolson
PMID66931
ABSTRACT
The effects of several commonly used detergents on the saccharide-binding activities of lectins were investigated using lectin-mediated agglutination of formalin-fixed erythrocytes and affinity chromatography of glycoproteins on columns of lectins immobilized on polyacrylic hydrazide-Sepharose. In the hemagglutination assays, Ricinus communis I (RCA1) and II (RCAII), concanavalin A (Con A), and the agglutinins from peanut (PNA), soybean (SBA), wheat germ (WGA), and Limulus polyphemus (LPA) were tested with several concentrations of switterionic, cationic, anionic, and nonionic detergents. It was found that increasing detergent concentrations eventually affected hemagglutination titers in both test and control samples, and the highest detergent concentrations not affecting lectin hemagglutinating activities were determined. The effects of detergents on specific binding of [3H]fetuin and asialo[3H]fetuin to and elution from columns of immobilized lectins were less severe when compared with lectins in solution, suggesting that the lectins are stabilized by covalent attachment to agarose beads. Nonionic detergents did not affect the binding efficiency of the immobilized lectins tested at concentrations used for membrane solubilization while cationic and zwitterionic detergents caused significant inhibition of Con A- and SBA-Sepharose activities. In sodium deoxycholate (greater than 1%) only RCAI-Sepharose retained its activity, whereas the activities of the other lectins were reduced dramatically. Low concentrations of sodium dodecyl sulfate (0.05%) inhibited only the activity of immobilized SBA, but at higher concentration (0.1%) and prolonged periods of incubation (16 h, 23 degrees C) most of the lectins were inactivated. These data are compared with previous reports on the use of detergents in lectin affinity chromatography, and the conditions for the optimal use of detergents are detailed.
Sigma-Aldrich
Concanavalin A from Canavalia ensiformis (Jack bean), agarose conjugate, Type VI, saline suspension
Pricing and availability is not currently available.