Skip to Content
We are planning system maintenance on January 25, 2025, from 1:00 PM to 4:00 PM CST. This will impact both web and offline transactions, including online orders, quotes, price and availability checks, and order status inquiries. We apologize for any inconvenience.
Merck
Inducible gene expression in mammalian cells and mice.
Methods in molecular biology (Clifton, N.J.) (2004-07-23)
Wilfried Weber, Martin Fussenegger
ABSTRACT
Inducible expression of desired transgenes in mammalian cells and animals is a current priority in basic and applied research, biopharmaceutical manufacturing, gene therapy, and tissue engineering, as well as in drug discovery. Among the most prominent human-compatible transgene control technologies are engineered promoter/transactivator configurations that adjust heterologous target gene transcription in response to clinically licensed antibiotics (tetracyclines, streptogramins, macrolides). In this chapter we provide a detailed case study on macrolide-inducible expression of the human model glycoprotein SEAP (human placental secreted alkaline phosphatase) in transgenic Chinese hamster ovary (CHO) cell cultures or following implantation of microencapsulated CHO cells into mice.