Merck
  • Dissecting phenotypic transitions in metastatic disease via photoconversion-based isolation.
Dissecting phenotypic transitions in metastatic disease via photoconversion-based isolation.
eLife (2021-02-24)
Yogev Sela, Jinyang Li, Paola Kuri, Allyson J Merrell, Ning Li, Chris Lengner, Pantelis Rompolas, Ben Z Stanger
ABSTRACT
Cancer patients often harbor occult metastases, a potential source of relapse that is targetable only through systemic therapy. Studies of this occult fraction have been limited by a lack of tools with which to isolate discrete cells on spatial grounds. We developed PIC-IT, a photoconversion-based isolation technique allowing efficient recovery of cell clusters of any size - including single-metastatic cells - which are largely inaccessible otherwise. In a murine pancreatic cancer model, transcriptional profiling of spontaneously arising microcolonies revealed phenotypic heterogeneity, functionally reduced propensity to proliferate and enrichment for an inflammatory-response phenotype associated with NF-κB/AP-1 signaling. Pharmacological inhibition of NF-κB depleted microcolonies but had no effect on macrometastases, suggesting microcolonies are particularly dependent on this pathway. PIC-IT thus enables systematic investigation of metastatic heterogeneity. Moreover, the technique can be applied to other biological systems in which isolation and characterization of spatially distinct cell populations is not currently feasible.
Sigma-Aldrich
Monoclonal Anti-phospho-Histone H3 (pSer28) antibody produced in rat, ~0.5 mg/mL, clone HTA28, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-p21 Antibody, clone 5G7, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells