Merck
  • Thermochemical formation of dioxins promoted by chromium chloride: In situ Cr- and Cl-XAFS analysis.
Thermochemical formation of dioxins promoted by chromium chloride: In situ Cr- and Cl-XAFS analysis.
Journal of hazardous materials (2020-01-19)
Mengmei Zhang, Takashi Fujimori, Kenji Shiota, Alfons Buekens, Kota Mukai, Yasuhiro Niwa, Xiaodong Li, Masaki Takaoka
ABSTRACT
Chromium is commonly found in the flue gases and ashes of Municipal Solid Waste Incineration. It has been reported as an active catalyst for the formation of polychlorinated dibenzo-p-dioxins and dibenzofurans (PCDD/F) during de novo tests, yet its specific mode of action has remained unclear. This study aims to identify the effects of chromium chloride on the formation of PCDD/F and other chloro-aromatics and to elucidate the underlying reaction mechanisms. A series of de novo tests, conducted over a wide range of temperature (from 250 to 550 °C) and for four different oxygen contents (0, 5, 10, 20 %), confirmed the promoting effect of CrCl3 on the PCDD/F formation. In situ X-ray Absorption Fine Structure (XAFS) spectroscopy was applied to investigate the behavior of CrCl3 during heating, describing the entire picture of CrCl3-promoted formation pathways of dioxins. The effect of oxygen was studied by measuring XAFS spectra on samples heated at different oxygen concentrations. According to these spectra, chromium compounds play two key roles during dioxins formation: (a) chlorinating carbon, using chlorine derived from conversion of CrCl3 into Cr2O3, and further oxidation to Cr(VI), and (b) facilitating oxidative destruction of the carbon matrix, while reducing Cr(VI) to Cr2O3.
Product Description
Carbon, tube, outside diameter 3.18 mm, length 150 mm, wall thickness 0.8 mm
Pricing and availability is not currently available.