Merck
  • Steady-state kinetic characterization of substrates and metal-ion specificities of the full-length and N-terminally truncated recombinant human methionine aminopeptidases (type 2).
Steady-state kinetic characterization of substrates and metal-ion specificities of the full-length and N-terminally truncated recombinant human methionine aminopeptidases (type 2).
Biochemistry (2001-08-29)
G Yang, R B Kirkpatrick, T Ho, G F Zhang, P H Liang, K O Johanson, D J Casper, M L Doyle, J P Marino, S K Thompson, W Chen, D G Tew, T D Meek
ABSTRACT
The steady-state kinetics of a full-length and truncated form of the type 2 human methionine aminopeptidase (hMetAP2) were analyzed by continuous monitoring of the amide bond cleavage of various peptide substrates and methionyl analogues of 7-amido-4-methylcoumarin (AMC) and p-nitroaniline (pNA), utilizing new fluorescence-based and absorbance-based assay substrates and a novel coupled-enzyme assay method. The most efficient substrates for hMetAP2 appeared to be peptides of three or more amino acids for which the values of k(cat)/K(m) were approximately 5 x 10(5) M(-1) min(-1). It was found that while the nature of the P1' residue of peptide substrates dictates the substrate specificity in the active site of hMetAP2, the P2' residue appears to play a key role in the kinetics of peptidolysis. The catalytic efficiency of dipeptide substrates was found to be at least 250-fold lower than those of the tripeptides. This substantially diminished catalytic efficiency of hMetAP2 observed with the alternative substrates MetAMC and MetpNA is almost entirely due to the reduction in the turnover rate (k(cat)), suggesting that cleavage of the amide bond is at least partially rate-limiting. The 107 N-terminal residues of hMetAP2 were not required for either the peptidolytic activity of the enzyme or its stability. Steady-state kinetic comparison and thermodynamic analyses of an N-terminally truncated form and full-length enzyme yielded essentially identical kinetic behavior and physical properties. Addition of exogenous Co(II) cation was found to significantly activate the full-length hMetAP2, while Zn(II) cation, on the other hand, was unable to activate hMetAP2 under any concentration that was tested.
Sigma-Aldrich
L-Methionine 7-amido-4-methylcoumarin trifluoroacetate salt, ≥99.0% (sum of enantiomers, HPLC)