Merck
  • Photoinduced Betaine Generation for Efficient Photothermal Energy Conversion.
Photoinduced Betaine Generation for Efficient Photothermal Energy Conversion.
Chemistry (Weinheim an der Bergstrasse, Germany) (2019-12-15)
Devika Sasikumar, Yuta Takano, Vasudevanpillai Biju
ABSTRACT
The conversion of solar energy to thermal, chemical, or electrical energy attracts great attention in chemistry and physics. There has been a considerable effort for the efficient extraction of photons throughout the entire solar spectrum. In this work light energy was efficiently harvested by using a long-lived betaine photogenerated from an acridinium-based electron donor-acceptor dyad. The photothermal energy-conversion efficiency of the dyad is significantly enhanced by simultaneous illumination with blue (420-440 nm) and yellow (>480 nm) light in comparison with the sum of the conversion efficiencies for individual illumination with blue or yellow light. The enhanced photothermal effect is due to the photogenerated betaine, which absorbs longer-wavelength light than the dyad, and thus the dyad-betaine combination is promising for efficient photothermal energy conversion. The mechanisms of betaine generation and energy conversion are discussed on the basis of steady-state and transient spectral measurements.
Product Description
Carbon, tube, outside diameter 3.18 mm, length 150 mm, wall thickness 0.8 mm
Pricing and availability is not currently available.