Merck
  • Polypyridyl Zinc(II)-Indomethacin Complexes with Potent Anti-Breast Cancer Stem Cell Activity.
Polypyridyl Zinc(II)-Indomethacin Complexes with Potent Anti-Breast Cancer Stem Cell Activity.
Molecules (Basel, Switzerland) (2018-09-06)
Tiffany K Rundstadler, Arvin Eskandari, Sarah M Norman, Kogularamanan Suntharalingam
ABSTRACT
Cancer stem cells (CSCs) are thought of as a clinically pertinent subpopulation of tumors, partly responsible for cancer relapse and metastasis. Research programs aimed at discovering anti-CSC agents have largely focused on biologics and purely organic molecules. Recently, we showed that a family of redox-active copper(II) complexes with phenanthroline-based ligands and nonsteroidal anti-inflammatory drugs (NSAIDs) such as indomethacin, are capable of potently and selectively killing breast CSCs. Herein we present analogous redox-inactive, zinc(II)-phenanthroline-indomethacin complexes with the ability to kill breast CSCs and bulk breast cancer cells with equal potency (in the submicro- or micromolar range). A single dose of the zinc(II) complexes could theoretically be administered to eliminate whole tumor populations. Excitingly, some of the zinc(II) complexes decrease the growth and viability of mammospheres to a comparable or higher degree than salinomycin, a compound known to effectively kill breast CSCs. As far as we are aware this is the first report to examine the anti-breast CSC activity of zinc(II)-containing compounds.
Sigma-Aldrich
5-Methyl-1,10-phenanthroline, ≥99%
Sigma-Aldrich
In Vitro Toxicology Assay Kit, Resazurin based
Sigma-Aldrich
1,10-Phenanthroline, ≥99%