
Thujone Standard Mixture

Synonym(s):
α,β-Thujone, (−)-1-Isopropyl-4-methylbicyclo[3.1.0]hexan-3-one
Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
Products
Brand
Boiling Point (°C)
Formula Weight
Format
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing