
SPhos Pd G2 ChemBeads

Synonym(s):
SPhos Pd G2, 2nd Generation SPhos Precatalyst, Chloro(2-dicyclohexylphosphino-2′,6′-dimethoxy-1,1′-biphenyl)[2-(2′-amino-1,1′-biphenyl)]palladium(II), SPhos-Pd-G2
Empirical Formula (Hill Notation):
C38H45ClNO2PPd
CAS Number:
Molecular Weight:
720.62
Products
Product Category
Feature
Formula Weight
Melting Point (°C)
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing