
Gly-Arg-Gly-Asp-Ser-Pro-Lys

Synonym(s):
GRGDSPK
Empirical Formula (Hill Notation):
C28H49N11O11
CAS Number:
Molecular Weight:
715.76
Products
Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing