
Epitetracycline hydrochloride

Synonym(s):
4-Epitetracycline hydrochloride, 4-epi-Tetracycline hydrochloride
Empirical Formula (Hill Notation):
C22H24N2O8 · HCl
CAS Number:
Molecular Weight:
480.90
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing