Merck
Biodegradation of dimethylterephthalate by mixed microbial cultures.
Acta microbiologica Bulgarica (1993-01-01)
L Tserovska, R Dimkov
ABSTRACT
The microbial association 189 was isolated from soil treated with dimethylterephthalate (DMT) for a long time. The dynamics of its growth, respectively the degradation of the terephthalate ester known as an environmental pollutant has been studied.
Product Description
Supelco
Dimethyl terephthalate, Standard for quantitative NMR, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dimethyl terephthalate, purum, ≥99.0% (GC)
Sigma-Aldrich
Dimethyl terephthalate, ReagentPlus®, ≥99%