Merck
E- and Z-p-methoxycinnamic acid from Aquilegia vulgaris.
Acta poloniae pharmaceutica (2004-12-04)
Wiesława Bylka
ABSTRACT
Two isomers E- and Z-p-methoxycinnamic acid were isolated from the leaves with stems of A. vulgaris L. and their structures were determined by 1HMR and 13C NMR spectral data.
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%