Merck
  • Functional analysis of acquired CD28 mutations identified in cutaneous T cell lymphoma.
Functional analysis of acquired CD28 mutations identified in cutaneous T cell lymphoma.
Cellular immunology (2017-07-18)
Grzegorz B Gmyrek, Jeanette Pingel, Jaehyuk Choi, Jonathan M Green
ABSTRACT
CD28 is the major costimulatory receptor on T cells regulating proliferation, survival and effector function. Acquired mutations in the extracellular domain of CD28 have been identified in patients with cutaneous T cell lymphoma, angioimmunoblastic T cell lymphoma and other T cell neoplasms, suggesting it may contribute to disease pathogenesis. We used a heterologous system in which mutant human CD28 was expressed on primary murine T cells deficient in CD28 to ascertain how specific mutations identified in a genetic screen of patients with cutaneous T cell lymphoma affected normal T cell function. All three mutant CD28 proteins examined enhanced CD28-dependent T cell proliferation and effector function. These data suggest that the mutant CD28 isoforms could accelerate tumor cell growth and increase tumor burden in affected patients. Interruption of CD28:ligand interactions may be an effective, targeted therapy for a subset of patients whose tumors bear the mutant CD28 receptor.
Sigma-Aldrich
N,O-Bis(trimethylsilyl)carbamate, ≥98.0% (T)