Merck
  • New 2-aryl-7,8-dimethoxy-3,4-dihydroisoquinolin-2-ium salts as potential antifungal agents: synthesis, bioactivity and structure-activity relationships.
New 2-aryl-7,8-dimethoxy-3,4-dihydroisoquinolin-2-ium salts as potential antifungal agents: synthesis, bioactivity and structure-activity relationships.
Scientific reports (2017-08-10)
Lifei Zhu, Bohang Zhou, Bingyu Zhang, Mingxuan Xu, Huiling Geng, Le Zhou
ABSTRACT
The title compounds can be considered as simple analogues of quaternary benzo[c]phenanthridine alkaloids (QBAs). In order to develop potent QBA-like antifungal agents, as our continuing study, a series of new title compounds were synthesized and evaluated for bioactivity against five plant pathogenic fungi by the mycelium growth rate method in this study. The SAR were also derived. The majority of the compounds showed good to excellent inhibition activity with average EC
Product Description
Supelco
Azoxystrobin, PESTANAL®, analytical standard