Merck
  • Photo-Fenton oxidation of 3-amino-5-methylisoxazole: a by-product from biological breakdown of some pharmaceutical compounds.
Photo-Fenton oxidation of 3-amino-5-methylisoxazole: a by-product from biological breakdown of some pharmaceutical compounds.
Environmental science and pollution research international (2015-11-12)
Bianca M Souza, Belisa A Marinho, Francisca C Moreira, Márcia W C Dezotti, Rui A R Boaventura, Vítor J P Vilar
ABSTRACT
The present study aims to assess the removal of 3-amino-5-methylisoxazole (AMI), a recalcitrant by-product resulting from the biological breakdown of some pharmaceuticals, applying a solar photo-Fenton process assisted by ferrioxalate complexes (SPFF) (Fe
Sigma-Aldrich
3-Amino-5-methylisoxazole, ≥97%