Merck
  • Inverted Micelle-in-Micelle Configuration in Cationic/Carbohydrate Surfactant Mixtures.
Inverted Micelle-in-Micelle Configuration in Cationic/Carbohydrate Surfactant Mixtures.
Chemphyschem : a European journal of chemical physics and physical chemistry (2016-10-28)
Saikat Das, Wenjin Xu, Hans-Joachim Lehmler, Anne-Frances Miller, Barbara L Knutson, Stephen E Rankin
ABSTRACT
Nuclear magnetic resonance is applied to investigate the relative positions and interactions between cationic and non-ionic carbohydrate-based surfactants in mixed micelles with D
Sigma-Aldrich
2-Naphthylboronic acid, ≥95.0%