Merck
[Antihypoxants in emergency medicine].
Anesteziologiia i reanimatologiia (1998-06-05)
A V Smirnov, B I Krivoruchko
ABSTRACT
Presents the results of many-year experimental studies, clinical trials, and medical use of drugs of a new pharmacological class: antihypoxants. Antihypoxants are perspective drugs of urgent medicine, which was demonstrated on various models (all types of hypoxia, myocardial, brain, renal, and liver ischemia, grave mechanical and thermal injuries, blood loss, etc.) and in patients with these states.
Sigma-Aldrich
2-Imino-4-thiobiuret, 99%