Merck
  • Pharmacological actions of a novel NO-independent guanylyl cyclase stimulator, BAY 41-8543: in vitro studies.
Pharmacological actions of a novel NO-independent guanylyl cyclase stimulator, BAY 41-8543: in vitro studies.
British journal of pharmacology (2002-01-30)
Johannes-Peter Stasch, Cristina Alonso-Alija, Heiner Apeler, Klaus Dembowsky, Achim Feurer, Torsten Minuth, Elisabeth Perzborn, Matthias Schramm, Alexander Straub
ABSTRACT
BAY 41-8543 is a novel, highly specific and so far the most potent NO-independent stimulator of sGC. Here we report the effects of BAY 41-8543 on the isolated enzyme, endothelial cells, platelets, isolated vessels and Langendorff heart preparation. BAY 41-8543 stimulates the recombinant sGC concentration-dependently from 0.0001 microM to 100 microM up to 92-fold. In combination, BAY 41-8543 and NO have synergistic effects over a wide range of concentrations. Similar results are shown in implying that BAY 41-8543 stimulates the sGC directly and furthermore makes the enzyme more sensitive to its endogenous activator NO. In vitro, BAY 41-8543 is a potent relaxing agent of aortas, saphenous arteries, coronary arteries and veins with IC(50)-values in the nM range. In the rat heart Langendorff preparation, BAY 41-8543 potently reduces coronary perfusion pressure from 10(-9) to 10(-6) g ml(-1) without any effect on left ventricular pressure and heart rate. BAY 41-8543 is effective even under nitrate tolerance conditions proved by the same vasorelaxing effect on aortic rings taken either from normal or nitrate-tolerant rats. BAY 41-8543 is a potent inhibitor of collagen-mediated aggregation in washed human platelets (IC(50)=0.09 microM). In plasma, BAY 41-8543 inhibits collagen-mediated aggregation better than ADP-induced aggregation, but has no effect on the thrombin pathway. BAY 41-8543 is also a potent direct stimulator of the cyclic GMP/PKG/VASP pathway in platelets and synergizes with NO over a wide range of concentrations. These results suggest that BAY 41-8543 is on the one hand an invaluable tool for studying sGC signaling in vitro and on the other hand its unique profile may offer a novel approach for treating cardiovascular diseases.
Sigma-Aldrich
BAY 41-8543, ≥98% (HPLC)