Merck
  • Angiotensin-converting enzyme inhibitor attenuates monocyte adhesion to vascular endothelium through modulation of intracellular zinc.
Angiotensin-converting enzyme inhibitor attenuates monocyte adhesion to vascular endothelium through modulation of intracellular zinc.
The Journal of pharmacology and experimental therapeutics (2007-09-20)
Chiari Kojima, Akio Kawakami, Takashi Takei, Kosaku Nitta, Masayuki Yoshida
ABSTRACT
To elucidate an anti-inflammatory role of angiotensin-converting enzyme inhibitors (ACEIs) in cardiovascular disease, we studied the effect of ACEIs in monocyte adhesion to endothelial cells and underlying molecular mechanisms. Treatment of human monocytic THP-1 cells with monocyte chemoattractant protein-1 (MCP-1; 100 ng/ml; 10 min) significantly increased their adhesion to human umbilical vein endothelial cells (HUVECs) under flow condition (P < 0.001). Preincubation of THP-1 cells with imidaprilat (50 nM; 4 h), an active metabolite of imidapril, reduced MCP-1-triggered THP-1 cell adhesion (P < 0.01). Similar effects were obtained with experiments using human peripheral monocytes (P < 0.05). MCP-1 activated protein kinase C (PKC)alpha in THP-1 cells, resulting in the up-regulation of alpha4 and beta2 integrin. Imidaprilat attenuated MCP-1-induced PKC activation and integrin up-regulation in THP-1 cells. Imidaprilat also inhibited THP-1 cell adhesion induced by phorbol 12-myristate 13-acetate (PMA), a potent PKC activator. In attempt to elucidate the mechanisms for the modulation of PKC activity by imidaprilat, we found that MCP-1 or PMA increased labile zinc in THP-1 cells, which was canceled by imidaprilat. Indeed, zinc/pyrithione activated PKC and increased THP-1 cell adhesion. Zinc chelator as well as PKC inhibitor inhibited these processes, suggesting the role for labile zinc in PKC activation and THP-1 cell adhesion. Imidaprilat attenuated zinc/pyrithione-induced PKC activation and THP-1 cell adhesion. These data suggest that ACEI reduces MCP-1 or PMA-triggered monocyte adhesion to activated HUVECs by modulating labile zinc in monocytes. Our findings may point out a novel anti-inflammatory mechanism of ACEIs in atherogenesis.
Sigma-Aldrich
Anti-Angiotensin Converting Enzyme Antibody, clone 9B9, clone 9B9, Chemicon®, from mouse