Merck
  • Asymmetric [4 + 3] cycloadditions between vinylcarbenoids and dienes: application to the total synthesis of the natural product (-)-5-epi-vibsanin E.
Asymmetric [4 + 3] cycloadditions between vinylcarbenoids and dienes: application to the total synthesis of the natural product (-)-5-epi-vibsanin E.
Journal of the American Chemical Society (2009-05-19)
Brett D Schwartz, Justin R Denton, Yajing Lian, Huw M L Davies, Craig M Williams
ABSTRACT
The total synthesis of (-)-5-epi-vibsanin E (2) has been achieved in 18 steps. The synthesis combines the rhodium-catalyzed [4 + 3] cycloaddition between a vinylcarbenoid and a diene to rapidly generate the tricyclic core with an effective end game strategy to introduce the remaining side-chains. The [4 + 3] cycloaddition occurs by a cyclopropanation to form a divinylcyclopropane followed by a Cope rearrangement to form a cycloheptadiene. The quaternary stereogenic center generated in the process can be obtained with high asymmetric induction when the reaction is catalyzed by the chiral dirhodium complex, Rh(2)(S-PTAD)(4).
Sigma-Aldrich
Rh2(R-PTAD)4
Sigma-Aldrich
Rh2(S-PTAD)4