
Voriconazole impurity D

Synonym(s):
(2S,3R)-2-(2,4-Difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, epi-Voriconazole
Empirical Formula (Hill Notation):
C16H14F3N5O
CAS Number:
Molecular Weight:
349.31
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing