Skip to Content
Merck
Flavoxate Related Compound B pharmaceutical secondary standard, certified reference material

Flavoxate Related Compound B

Flavoxate Related Compound B pharmaceutical secondary standard, certified reference material
Synonym(s):
Methyl 3-methyl-4-oxo-2-phenyl-4H-benzopyrane-8-carboxylate, 3-Methylflavone-8-carboxylic acid methyl ester
Empirical Formula (Hill Notation):
C18H14O4
CAS Number:
Molecular Weight:
294.30

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade