
Ciprofloxacin Related Compound A

Synonym(s):
7-Chloro-1-cyclopropyl-4-oxo-6-(piperazin-1-yl)-1,4-dihydroquinoline-3-carboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C17H18ClN3O3 · HCl
CAS Number:
Molecular Weight:
384.26
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2