
Almotriptan Related Compound D

Synonym(s):
N,N-Dimethyl-5-[(1-pyrrolidinylsulfonyl)methyl]-1H-indole-3-ethanamine N-oxide hydrochloride, 1-[({3-[2-(Dimethylamino)ethyl]indol-5-yl}methyl)sulfonyl]pyrrolidine N-oxide
Empirical Formula (Hill Notation):
C17H25N3O3S · HCl
CAS Number:
Molecular Weight:
387.92
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2