Skip to Content
Merck
2-Pyrrolidinone purum, ≥98.0% (GC)

2-Pyrrolidinone

2-Pyrrolidinone purum, ≥98.0% (GC)
Synonym(s):
2-Pyrrolidone, Butyrolactam
Empirical Formula (Hill Notation):
C4H7NO
CAS Number:
Molecular Weight:
85.10
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing