![1,4-Bis(trimethylsilyl)butadiyne 98%, stable crystalline form of butadiyne](/deepweb/assets/sigmaaldrich/product/structures/410/107/bf8ce167-1f68-43af-942d-ddfcbe3e5c4a/640/bf8ce167-1f68-43af-942d-ddfcbe3e5c4a.png)
1,4-Bis(trimethylsilyl)butadiyne
![1,4-Bis(trimethylsilyl)butadiyne 98%, stable crystalline form of butadiyne](/deepweb/assets/sigmaaldrich/product/structures/410/107/bf8ce167-1f68-43af-942d-ddfcbe3e5c4a/640/bf8ce167-1f68-43af-942d-ddfcbe3e5c4a.png)
Synonym(s):
1,4-Bis(trimethylsilyl)-1,3-butadiyne, BTMSBD
Linear Formula:
(CH3)3SiC≡CC≡CSi(CH3)3
CAS Number:
Molecular Weight:
194.42
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing