- Simple and reliable method to precipitate proteins from bacterial culture supernatant.
Simple and reliable method to precipitate proteins from bacterial culture supernatant.
Applied and environmental microbiology (2004-01-09)
Randolph B Caldwell, Claus T Lattemann
PMID14711696
RESUMEN
A simple and reliable method for precipitating protein from bacterial culture supernatants based on a pyrogallol red-molybdate-methanol (PRMM) protocol has been developed and applied for the analysis of proteins secreted by a bacterial type III secretion system. PRMM-based precipitation has been shown to be more efficient and robust than are conventional protocols.