Merck
  • The possible involvement of 3-methoxy-2(5H)-furanone in the etiology of Narthecium asiaticum maxim. associated nephrotoxicity in cattle.
The possible involvement of 3-methoxy-2(5H)-furanone in the etiology of Narthecium asiaticum maxim. associated nephrotoxicity in cattle.
Natural toxins (2000-12-21)
A Flåøyen, T L Torgersen, W Langseth
ABSTRACT
Two samples of Narthecium asiaticum Maxim leaves collected in Japan were found to contain 103 microg(-1) and 160 microg g(-1) dry matter of 3-methoxy-2(5H)-furanone respectively. 3-Methoxy-2(5h)-furanone was suggested to be the toxic principle of N. asiaticum causing nephrotoxicity in cattle in Japan. Two other furanones, which are thought to be non-toxic, were also isolated from the two samples. These were 4-methoxy-2(5H)-furanone and 5-hydroxy-4-methoxy-2(5H)-furanone.
Sigma-Aldrich
4-Methoxy-2(5H)-furanone, 99%