Merck
  • Comparison of the Retention and Separation Selectivity of Aromatic Hydrocarbons with Polar Groups in RP-HPLC Systems with Different Stationary Phases and Eluents.
Comparison of the Retention and Separation Selectivity of Aromatic Hydrocarbons with Polar Groups in RP-HPLC Systems with Different Stationary Phases and Eluents.
Molecules (Basel, Switzerland) (2020-11-04)
Anna Klimek-Turek, Beata Misiołek, Tadeusz H Dzido
ABSTRACT
In this manuscript, the retention of aromatic hydrocarbons with polar groups has been compared for systems with various nonpolar columns of the types from C3 to C18 and different mobile phases composed of methanol, acetonitrile, or tetrahydrofuran as modifiers. The selectivity separation of the solutes in systems with different adsorbents, when one eluent modifier is swapped by another, has been explained, taking into account molecular interactions of the solutes with components of the stationary phase region (i.e., extracted modifier depending on the chain length of the stationary phase).
Sigma-Aldrich
1,7-Dihydroxynaphthalene, 97%
Supelco
Methyl benzoate, analytical standard