Merck
  • Rationale and design of the expanded combination of evolocumab plus empagliflozin in diabetes: EXCEED-BHS3 trial.
Rationale and design of the expanded combination of evolocumab plus empagliflozin in diabetes: EXCEED-BHS3 trial.
Therapeutic advances in chronic disease (2020-10-17)
Ikaro Breder, Jessica Cunha Breder, Isabella Bonilha, Daniel B Munhoz, Sheila T Kimura Medorima, Daniela C Oliveira, Helison R do Carmo, Camila Moreira, Anatol Kontush, Francesca Zimetti, Ilaria Zanotti, Luiz Sergio F Carvalho, Wilson Nadruz, Elza Muscelli, Thiago Quinaglia, Andrei C Sposito
ABSTRACT
Patients with type 2 diabetes mellitus (T2DM) remain at increased cardiovascular residual risk and endothelial dysfunction, even after optimizing metabolic control and treatment by sodium-glucose-2 transporter inhibitors (SGLT2-is). The present study was based on the hypothesis that proprotein convertase subtilisin/kexin 9 inhibitor (PCSK9i) therapy may mitigate endothelial dysfunction in T2DM patients who are on regular treatment by SGLT2-i. The EXCEED-BHS3 is a prospective, single-center, investigator-blinded, open-label, randomized clinical trial. Participants (n = 110) will be randomized (1:1) to either empagliflozin 25 mg/day alone or empagliflozin 25 mg/day plus evolocumab 140 mg every 2 weeks in addition to optimal medical care. The primary endpoint was defined as the change in the 1-min flow-mediated dilation (FMD) after 16 weeks of treatment. The secondary endpoint is the FMD change after ischemia/reperfusion injury protocol (reserve FMD) after 16 weeks of treatment. Exploratory outcomes comprise the change in FMD and reserve FMD after 8 weeks of treatment and the change after 16 weeks of treatment in the following parameters: plasma levels of nitric oxide, vascular cell adhesion molecule-1 and isoprostane, high-density lipoprotein (HDL) and low-density lipoprotein subfractions profile, HDL function, blood pressure, body mass index, waist circumference and adipokines. This will be the first study to evaluate the add-on effect of PCSK9i on endothelial function of T2DM patients under regular use of empagliflozin. ClinicalTrials.gov identifier: NCT03932721.