Merck
  • Proton myo-inositol cotransporter is a novel γ-secretase associated protein that regulates Aβ production without affecting Notch cleavage.
Proton myo-inositol cotransporter is a novel γ-secretase associated protein that regulates Aβ production without affecting Notch cleavage.
The FEBS journal (2015-06-23)
Yasuhiro Teranishi, Mitsuhiro Inoue, Natsuko Goto Yamamoto, Takahiro Kihara, Birgitta Wiehager, Taizo Ishikawa, Bengt Winblad, Sophia Schedin-Weiss, Susanne Frykman, Lars O Tjernberg
ABSTRACT
γ-Secretase is a transmembrane protease complex that is responsible for the processing of a multitude of type 1 transmembrane proteins, including the amyloid precursor protein and Notch. γ-Secretase processing of amyloid precursor protein results in the release of the amyloid β-peptide (Aβ), which is involved in the pathogenesis in Alzheimer's disease. Processing of Notch leads to the release of its intracellular domain, which is important for cell development. γ-Secretase associated proteins (GSAPs) could be of importance for substrate selection, and we have previously shown that affinity purification of γ-secretase in combination with mass spectrometry can be used for finding such proteins. In the present study, we used this methodology to screen for novel GSAPs from human brain, and studied their effect on Aβ production in a comprehensive gene knockdown approach. Silencing of probable phospholipid-transporting ATPase IIA, brain-derived neurotrophic factor/neurotrophin-3 growth factor receptor precursor and proton myo-inositol cotransporter (SLC2A13) showed a clear reduction of Aβ and these proteins were selected for further studies on Aβ production and Notch cleavage using small interfering RNA-mediated gene silencing, as well as an overexpression approach. Silencing of these reduced Aβ secretion in a small interfering RNA dose-dependent manner. Interestingly, SLC2A13 had a lower effect on Notch processing. Furthermore, overexpression of SLC2A13 increased Aβ40 generation. Finally, the interaction between γ-secretase and SLC2A13 was confirmed using immunoprecipitation and a proximity ligation assay. In summary, SLC2A13 was identified as a novel GSAP that regulates Aβ production without affecting Notch cleavage. We suggest that SLC2A13 could be a target for Aβ lowering therapy aimed at treating Alzheimer's disease.
Sigma-Aldrich
Monoclonal Anti-NTRK2 antibody produced in mouse, clone 4D3-F10, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Nicastrin Antibody, ascites fluid, clone 9C3, Chemicon®
Sigma-Aldrich
Anti-Nicastrin antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution